Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615388 | ||||||||||||||||||||
Literature Links: |
ADAT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAT2 - adenosine deaminase, tRNA specific 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286259.1 | 5650 | UTR 3 | NP_001273188.1 | |||
NM_182503.2 | 5650 | UTR 3 | NP_872309.2 | |||
XM_011535439.2 | 5650 | UTR 3 | XP_011533741.1 | |||
XM_011535440.1 | 5650 | UTR 3 | XP_011533742.1 | |||
XM_011535443.1 | 5650 | Intron | XP_011533745.1 | |||
XM_011535444.2 | 5650 | UTR 3 | XP_011533746.1 | |||
XM_017010260.1 | 5650 | UTR 3 | XP_016865749.1 | |||
XM_017010261.1 | 5650 | UTR 3 | XP_016865750.1 | |||
XM_017010262.1 | 5650 | UTR 3 | XP_016865751.1 | |||
XM_017010263.1 | 5650 | UTR 3 | XP_016865752.1 |