Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMTC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMTC4 - transmembrane and tetratricopeptide repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079669.2 | 3840 | UTR 3 | NP_001073137.1 | |||
NM_001286453.1 | 3840 | UTR 3 | NP_001273382.1 | |||
NM_032813.3 | 3840 | UTR 3 | NP_116202.2 | |||
XM_011521121.2 | 3840 | Intron | XP_011519423.1 | |||
XM_011521122.2 | 3840 | Intron | XP_011519424.1 | |||
XM_011521123.2 | 3840 | Intron | XP_011519425.1 | |||
XM_011521124.2 | 3840 | Intron | XP_011519426.1 | |||
XM_011521125.1 | 3840 | Intron | XP_011519427.1 | |||
XM_017020796.1 | 3840 | Intron | XP_016876285.1 | |||
XM_017020797.1 | 3840 | Intron | XP_016876286.1 | |||
XM_017020798.1 | 3840 | Intron | XP_016876287.1 | |||
XM_017020799.1 | 3840 | Intron | XP_016876288.1 | |||
XM_017020800.1 | 3840 | Intron | XP_016876289.1 |