Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 314997 | ||||||||||||||||||||
Literature Links: |
ZNF75D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF75D - zinc finger protein 75D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185063.2 | 4023 | Intron | NP_001171992.1 | |||
NM_007131.4 | 4023 | UTR 3 | NP_009062.2 | |||
XM_005262469.1 | 4023 | UTR 3 | XP_005262526.1 | |||
XM_005262471.3 | 4023 | UTR 3 | XP_005262528.1 | |||
XM_011531391.1 | 4023 | UTR 3 | XP_011529693.1 | |||
XM_011531395.2 | 4023 | UTR 3 | XP_011529697.1 | |||
XM_011531396.2 | 4023 | UTR 3 | XP_011529698.1 | |||
XM_011531397.2 | 4023 | UTR 3 | XP_011529699.1 | |||
XM_017029818.1 | 4023 | UTR 3 | XP_016885307.1 | |||
XM_017029819.1 | 4023 | UTR 3 | XP_016885308.1 | |||
XM_017029820.1 | 4023 | UTR 3 | XP_016885309.1 |