Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGGAAATGTTGAGAGAATTACC[A/G]AAACGTGACAGCAACACGTAGTGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603526 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTA1 - metastasis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203258.1 | Intron | NP_001190187.1 | ||||
NM_004689.3 | Intron | NP_004680.2 | ||||
XM_011537305.1 | Intron | XP_011535607.1 | ||||
XM_011537306.1 | Intron | XP_011535608.1 | ||||
XM_011537307.1 | Intron | XP_011535609.1 | ||||
XM_011537308.2 | Intron | XP_011535610.1 | ||||
XM_011537309.1 | Intron | XP_011535611.1 | ||||
XM_011537310.1 | Intron | XP_011535612.1 | ||||
XM_017021752.1 | Intron | XP_016877241.1 | ||||
XM_017021753.1 | Intron | XP_016877242.1 | ||||
XM_017021754.1 | Intron | XP_016877243.1 | ||||
XM_017021755.1 | Intron | XP_016877244.1 | ||||
XM_017021756.1 | Intron | XP_016877245.1 | ||||
XM_017021757.1 | Intron | XP_016877246.1 | ||||
XM_017021758.1 | Intron | XP_016877247.1 | ||||
XM_017021759.1 | Intron | XP_016877248.1 |
Set Membership: |
HapMap |