Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCATAGGACGGGTTGGATACCA[C/T]GTATGTAGTTTCTGTGCCAGCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607852 | ||||||||||||||||||||||||||||||||
Literature Links: |
NKD2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
NKD2 - naked cuticle homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271082.1 | Intron | NP_001258011.1 | ||||
NM_033120.3 | Intron | NP_149111.1 | ||||
XM_005248382.3 | Intron | XP_005248439.1 | ||||
XM_005248383.3 | Intron | XP_005248440.1 | ||||
XM_011514150.2 | Intron | XP_011512452.1 | ||||
XM_017010000.1 | Intron | XP_016865489.1 |