Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610178 MIM: 607384 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0586 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA0586 - KIAA0586 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM9 - translocase of inner mitochondrial membrane 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304485.1 | Intron | NP_001291414.1 | ||||
NM_001304486.1 | Intron | NP_001291415.1 | ||||
NM_001304487.1 | Intron | NP_001291416.1 | ||||
NM_001304488.1 | Intron | NP_001291417.1 | ||||
NM_001304489.1 | Intron | NP_001291418.1 | ||||
NM_001304490.1 | Intron | NP_001291419.1 | ||||
NM_001304491.1 | Intron | NP_001291420.1 | ||||
NM_012460.3 | Intron | NP_036592.1 |
TOMM20L - translocase of outer mitochondrial membrane 20 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |