Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607165 | ||||||||||||||||||||
Literature Links: |
SERINC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERINC3 - serine incorporator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006811.3 | 2018 | UTR 3 | NP_006802.1 | |||
NM_198941.2 | 2018 | Intron | NP_945179.1 | |||
XM_017027601.1 | 2018 | UTR 3 | XP_016883090.1 |
TTPAL - alpha tocopherol transfer protein like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |