Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604866 MIM: 607491 | ||||||||||||||||||||
Literature Links: |
PLAGL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLAGL2 - PLAG1 like zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002657.3 | Intron | NP_002648.1 | ||||
XM_005260436.3 | Intron | XP_005260493.1 | ||||
XM_011528863.2 | Intron | XP_011527165.1 | ||||
XM_011528864.2 | Intron | XP_011527166.1 |
POFUT1 - protein O-fucosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPY26P - testis specific protein, Y-linked 26, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |