Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATTTTGAATTTTTTTTTTTACC[C/G]CAGGAACACAGATTCAAGTTGCTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194555 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100379224 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100379224 - uncharacterized LOC100379224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF224 - zinc finger protein 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF225 - zinc finger protein 225 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321685.1 | Intron | NP_001308614.1 | ||||
NM_013362.3 | Intron | NP_037494.2 | ||||
XM_005259223.3 | Intron | XP_005259280.1 | ||||
XM_011527285.2 | Intron | XP_011525587.1 | ||||
XM_011527286.2 | Intron | XP_011525588.1 |
Set Membership: |
HapMap |