Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300654 MIM: 300517 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAAH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAAH2 - fatty acid amide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIN2B - spindlin family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006681.1 | Intron | NP_001006682.1 | ||||
NM_001006682.1 | Intron | NP_001006683.1 | ||||
NM_001006683.1 | Intron | NP_001006684.1 | ||||
NM_001282461.1 | Intron | NP_001269390.1 | ||||
NM_001282462.1 | Intron | NP_001269391.1 | ||||
XM_005262008.4 | Intron | XP_005262065.2 | ||||
XM_005262009.4 | Intron | XP_005262066.2 | ||||
XM_005262010.1 | Intron | XP_005262067.1 | ||||
XM_011530788.2 | Intron | XP_011529090.1 | ||||
XM_011530789.2 | Intron | XP_011529091.1 | ||||
XM_017029552.1 | Intron | XP_016885041.1 |