Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609434 | |||||||||||||||||||||||
Literature Links: |
LUC7L3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LUC7L3 - LUC7 like 3 pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006107.3 | Intron | NP_006098.2 | ||||
NM_016424.4 | Intron | NP_057508.2 | ||||
XM_005257448.1 | Intron | XP_005257505.1 | ||||
XM_005257449.2 | Intron | XP_005257506.1 | ||||
XM_005257452.1 | Intron | XP_005257509.1 | ||||
XM_006721943.2 | Intron | XP_006722006.1 |