Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610305 MIM: 601607 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DERL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DERL3 - derlin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002862.2 | Intron | NP_001002862.1 | ||||
NM_001135751.1 | Intron | NP_001129223.1 | ||||
NM_198440.3 | Intron | NP_940842.2 | ||||
XM_011530505.2 | Intron | XP_011528807.1 | ||||
XM_011530506.2 | Intron | XP_011528808.1 | ||||
XM_017029078.1 | Intron | XP_016884567.1 | ||||
XM_017029079.1 | Intron | XP_016884568.1 | ||||
XM_017029080.1 | Intron | XP_016884569.1 | ||||
XM_017029081.1 | Intron | XP_016884570.1 | ||||
XM_017029082.1 | Intron | XP_016884571.1 |
SMARCB1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007468.2 | Intron | NP_001007469.1 | ||||
NM_001317946.1 | Intron | NP_001304875.1 | ||||
NM_003073.4 | Intron | NP_003064.2 | ||||
XM_011530345.2 | Intron | XP_011528647.1 |
Set Membership: |
HapMap |