Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTAAGAAAATTAGAGAAATGGAG[A/T]GATCATGTGTATGGTGTTACGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606783 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLEC12B - C-type lectin domain family 12 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129998.2 | Intron | NP_001123470.1 | ||||
NM_001319241.1 | Intron | NP_001306170.1 | ||||
NM_001319242.1 | Intron | NP_001306171.1 | ||||
NM_205852.3 | Intron | NP_995324.2 | ||||
XM_011520658.2 | Intron | XP_011518960.1 | ||||
XM_011520663.2 | Intron | XP_011518965.1 | ||||
XM_017019295.1 | Intron | XP_016874784.1 |
CLEC1B - C-type lectin domain family 1 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC102724020 - uncharacterized LOC102724020 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |