Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCCAGGTTCAAATGATTCTCC[C/T]ATGTCAGCCTTTCAAGTAACTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194543 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF69 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF69 - zinc finger protein 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321163.1 | Intron | NP_001308092.1 | ||||
NM_021915.3 | Intron | NP_068734.1 | ||||
XM_006722873.3 | Intron | XP_006722936.1 | ||||
XM_006722874.3 | Intron | XP_006722937.1 | ||||
XM_017027231.1 | Intron | XP_016882720.1 |