Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTACTTAAGAACAACCCCCCT[C/T]CACCTCCCAACCAGTAGTACACAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605325 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP3A5 - cytochrome P450 family 3 subfamily A member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000777.4 | Intron | NP_000768.1 | ||||
NM_001190484.2 | Intron | NP_001177413.1 | ||||
NM_001291829.1 | Intron | NP_001278758.1 | ||||
NM_001291830.1 | Intron | NP_001278759.1 | ||||
XM_006715859.2 | Intron | XP_006715922.1 | ||||
XM_011515843.1 | Intron | XP_011514145.1 | ||||
XM_017011781.1 | Intron | XP_016867270.1 | ||||
XM_017011782.1 | Intron | XP_016867271.1 |
ZSCAN25 - zinc finger and SCAN domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145115.2 | Intron | NP_660090.2 | ||||
XM_005250194.2 | Intron | XP_005250251.1 | ||||
XM_011515905.2 | Intron | XP_011514207.1 | ||||
XM_011515906.2 | Intron | XP_011514208.1 | ||||
XM_011515907.2 | Intron | XP_011514209.1 | ||||
XM_011515908.2 | Intron | XP_011514210.1 | ||||
XM_011515909.2 | Intron | XP_011514211.1 | ||||
XM_011515910.2 | Intron | XP_011514212.1 | ||||
XM_017011824.1 | Intron | XP_016867313.1 | ||||
XM_017011825.1 | Intron | XP_016867314.1 | ||||
XM_017011826.1 | Intron | XP_016867315.1 | ||||
XM_017011827.1 | Intron | XP_016867316.1 |
Set Membership: |
HapMap |