Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACAGAAAGAAAAGTAAAAGAGA[C/G]GTTGAAAGATCAAACTAACCAGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611649 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM188A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM188A - family with sequence similarity 188 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318330.1 | 2587 | Intron | NP_001305259.1 | |||
NM_024948.3 | 2587 | Intron | NP_079224.1 | |||
XM_005252600.2 | 2587 | Intron | XP_005252657.1 | |||
XM_005252602.2 | 2587 | Intron | XP_005252659.1 | |||
XM_006717508.2 | 2587 | Intron | XP_006717571.1 | |||
XM_011519690.2 | 2587 | UTR 3 | XP_011517992.1 | |||
XM_011519691.2 | 2587 | Intron | XP_011517993.1 | |||
XM_011519693.1 | 2587 | Intron | XP_011517995.1 | |||
XM_011519694.1 | 2587 | Intron | XP_011517996.1 | |||
XM_017016671.1 | 2587 | UTR 3 | XP_016872160.1 | |||
XM_017016672.1 | 2587 | Intron | XP_016872161.1 | |||
XM_017016673.1 | 2587 | Intron | XP_016872162.1 | |||
XM_017016674.1 | 2587 | Intron | XP_016872163.1 | |||
XM_017016675.1 | 2587 | Intron | XP_016872164.1 | |||
XM_017016676.1 | 2587 | Intron | XP_016872165.1 |