Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGAAATACTGAAGGGACACGT[C/T]TTCATGAAGCAGATCAAAATGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603760 MIM: 609721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HUS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HUS1 - HUS1 checkpoint clamp component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004507.3 | 909 | Missense Mutation | AAC,GAC | N,D 269 | NP_004498.1 | |
XM_017012115.1 | 909 | Missense Mutation | AAC,GAC | N,D 248 | XP_016867604.1 |
PKD1L1 - polycystin 1 like 1, transient receptor potential channel interacting | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |