Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605414 MIM: 602373 MIM: 611011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA7 - ATP binding cassette subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNN2 - calponin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303499.1 | Intron | NP_001290428.1 | ||||
NM_001303501.1 | Intron | NP_001290430.1 | ||||
NM_004368.3 | Intron | NP_004359.1 | ||||
NM_201277.2 | Intron | NP_958434.1 |
RNU6-2 - RNA, U6 small nuclear 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM259 - transmembrane protein 259 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |