Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTCTTGAATGTTTTGTTCCCGA[C/T]GAACTCAAGGAGCGAGACCTCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2AK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2AK3 - eukaryotic translation initiation factor 2 alpha kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313915.1 | 3593 | Silent Mutation | TCA,TCG | S,S 946 | NP_001300844.1 | |
NM_004836.6 | 3593 | Silent Mutation | TCA,TCG | S,S 1097 | NP_004827.4 | |
XM_017005376.1 | 3593 | Silent Mutation | TCA,TCG | S,S 869 | XP_016860865.1 |
LOC101928371 - uncharacterized LOC101928371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |