Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTATCTGTTCCCTATGGTTTCAA[A/T]CTAACTAAAACCTTATCAATTTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605655 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXL5 - F-box and leucine rich repeat protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193534.1 | Intron | NP_001180463.1 | ||||
NM_001193535.1 | Intron | NP_001180464.1 | ||||
NM_012161.3 | Intron | NP_036293.1 | ||||
XM_006713959.3 | Intron | XP_006714022.1 | ||||
XM_011513831.2 | Intron | XP_011512133.1 | ||||
XM_011513832.2 | Intron | XP_011512134.1 | ||||
XM_011513833.2 | Intron | XP_011512135.1 | ||||
XM_017008019.1 | Intron | XP_016863508.1 |