Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300522 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQSEC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IQSEC2 - IQ motif and Sec7 domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111125.2 | Intron | NP_001104595.1 | ||||
NM_001243197.1 | Intron | NP_001230126.1 | ||||
NM_015075.1 | Intron | NP_055890.1 | ||||
XM_006724579.2 | Intron | XP_006724642.1 | ||||
XM_006724580.3 | Intron | XP_006724643.1 | ||||
XM_006724581.3 | Intron | XP_006724644.1 | ||||
XM_006724582.3 | Intron | XP_006724645.1 | ||||
XM_006724583.3 | Intron | XP_006724646.1 | ||||
XM_006724584.2 | Intron | XP_006724647.1 | ||||
XM_011530773.2 | Intron | XP_011529075.1 | ||||
XM_011530774.2 | Intron | XP_011529076.1 | ||||
XM_011530776.1 | Intron | XP_011529078.1 | ||||
XM_011530777.1 | Intron | XP_011529079.1 | ||||
XM_017029359.1 | Intron | XP_016884848.1 | ||||
XM_017029360.1 | Intron | XP_016884849.1 |
Set Membership: |
HapMap |