Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610702 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C20orf27 - chromosome 20 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPA12B - heat shock protein family A (Hsp70) member 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197327.1 | Intron | NP_001184256.1 | ||||
NM_001318322.1 | Intron | NP_001305251.1 | ||||
NM_052970.4 | Intron | NP_443202.3 | ||||
XM_011529151.2 | Intron | XP_011527453.1 | ||||
XM_017027632.1 | Intron | XP_016883121.1 | ||||
XM_017027633.1 | Intron | XP_016883122.1 | ||||
XM_017027634.1 | Intron | XP_016883123.1 | ||||
XM_017027635.1 | Intron | XP_016883124.1 |
Set Membership: |
HapMap |