Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTCTCACCCACTTCCAATACA[G/T]CTTCCTTGCTGTCTCCAGCTTGAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 217030 MIM: 611652 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFI PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFI - complement factor I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000204.4 | Intron | NP_000195.2 | ||||
NM_001318057.1 | Intron | NP_001304986.1 | ||||
XM_005262976.1 | Intron | XP_005263033.1 | ||||
XM_006714209.1 | Intron | XP_006714272.1 | ||||
XM_006714210.3 | Intron | XP_006714273.1 | ||||
XM_011531920.1 | Intron | XP_011530222.1 | ||||
XM_017008164.1 | Intron | XP_016863653.1 | ||||
XM_017008165.1 | Intron | XP_016863654.1 | ||||
XM_017008166.1 | Intron | XP_016863655.1 |
PLA2G12A - phospholipase A2 group XIIA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |