Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615388 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAT2 - adenosine deaminase, tRNA specific 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286259.1 | 3914 | UTR 3 | NP_001273188.1 | |||
NM_182503.2 | 3914 | UTR 3 | NP_872309.2 | |||
XM_011535439.2 | 3914 | UTR 3 | XP_011533741.1 | |||
XM_011535440.1 | 3914 | UTR 3 | XP_011533742.1 | |||
XM_011535443.1 | 3914 | Intron | XP_011533745.1 | |||
XM_011535444.2 | 3914 | UTR 3 | XP_011533746.1 | |||
XM_017010260.1 | 3914 | UTR 3 | XP_016865749.1 | |||
XM_017010261.1 | 3914 | UTR 3 | XP_016865750.1 | |||
XM_017010262.1 | 3914 | UTR 3 | XP_016865751.1 | |||
XM_017010263.1 | 3914 | UTR 3 | XP_016865752.1 |
Set Membership: |
HapMap |