Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133520 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ERCC4 - ERCC excision repair 4, endonuclease catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005236.2 | 4071 | UTR 3 | NP_005227.1 | |||
XM_011522424.2 | 4071 | UTR 3 | XP_011520726.1 | |||
XM_011522427.1 | 4071 | UTR 3 | XP_011520729.1 | |||
XM_017023043.1 | 4071 | Intron | XP_016878532.1 |
Set Membership: |
HapMap |