Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602539 MIM: 608626 | ||||||||||||||||||||
Literature Links: |
LIMD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIMD2 - LIM domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC729683 - uncharacterized LOC729683 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K3 - mitogen-activated protein kinase kinase kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRADA - STE20-related kinase adaptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003786.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 347 | NP_001003786.1 | |
NM_001003787.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 384 | NP_001003787.1 | |
NM_001003788.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 326 | NP_001003788.1 | |
NM_001165969.1 | 1246 | UTR 3 | NP_001159441.1 | |||
NM_001165970.1 | 1246 | UTR 3 | NP_001159442.1 | |||
NM_153335.5 | 1246 | UTR 3 | NP_699166.2 | |||
XM_005257797.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 376 | XP_005257854.1 | |
XM_005257798.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 355 | XP_005257855.1 | |
XM_005257799.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 326 | XP_005257856.1 | |
XM_005257800.2 | 1246 | UTR 3 | XP_005257857.1 | |||
XM_005257801.4 | 1246 | Intron | XP_005257858.1 | |||
XM_005257803.4 | 1246 | Intron | XP_005257860.1 | |||
XM_011525466.2 | 1246 | Silent Mutation | AGC,AGT | S,S 369 | XP_011523768.1 | |
XM_011525467.2 | 1246 | Nonsense Mutation | CGA,TGA | R,* 349 | XP_011523769.1 | |
XM_017025312.1 | 1246 | UTR 3 | XP_016880801.1 | |||
XM_017025313.1 | 1246 | UTR 3 | XP_016880802.1 | |||
XM_017025314.1 | 1246 | Intron | XP_016880803.1 |