Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEAL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCEAL4 - transcription elongation factor A like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006935.2 | Intron | NP_001006936.1 | ||||
NM_001006937.2 | Intron | NP_001006938.1 | ||||
NM_001300901.1 | Intron | NP_001287830.1 | ||||
NM_001305840.1 | Intron | NP_001292769.1 | ||||
NM_001305841.1 | Intron | NP_001292770.1 | ||||
NM_001305842.1 | Intron | NP_001292771.1 | ||||
NM_024863.5 | Intron | NP_079139.4 | ||||
XM_017029856.1 | Intron | XP_016885345.1 | ||||
XM_017029857.1 | Intron | XP_016885346.1 |
Set Membership: |
HapMap |