Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613842 MIM: 605811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GZF1 - GDNF inducible zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317012.1 | 148 | Silent Mutation | CTA,TTA | L,L 17 | NP_001303941.1 | |
NM_001317019.1 | 148 | Silent Mutation | CTA,TTA | L,L 17 | NP_001303948.1 | |
NM_022482.4 | 148 | Silent Mutation | CTA,TTA | L,L 17 | NP_071927.1 | |
XM_005260800.2 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_005260857.1 | |
XM_005260802.3 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_005260859.1 | |
XM_011529321.2 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_011527623.1 | |
XM_011529322.1 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_011527624.1 | |
XM_017028017.1 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_016883506.1 | |
XM_017028018.1 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_016883507.1 | |
XM_017028019.1 | 148 | Silent Mutation | CTA,TTA | L,L 17 | XP_016883508.1 |
LINC01431 - long intergenic non-protein coding RNA 1431 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NXT1 - nuclear transport factor 2 like export factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |