Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604696 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKAP11 - A-kinase anchoring protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016248.3 | Intron | NP_057332.1 | ||||
XM_005266247.1 | Intron | XP_005266304.1 | ||||
XM_005266248.3 | Intron | XP_005266305.1 | ||||
XM_005266249.3 | Intron | XP_005266306.1 | ||||
XM_005266250.3 | Intron | XP_005266307.1 | ||||
XM_011534903.2 | Intron | XP_011533205.1 | ||||
XM_011534904.1 | Intron | XP_011533206.1 | ||||
XM_011534905.1 | Intron | XP_011533207.1 | ||||
XM_011534906.2 | Intron | XP_011533208.1 | ||||
XM_017020381.1 | Intron | XP_016875870.1 | ||||
XM_017020382.1 | Intron | XP_016875871.1 | ||||
XM_017020383.1 | Intron | XP_016875872.1 |
Set Membership: |
HapMap |