Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DZANK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DZANK1 - double zinc ribbon and ankyrin repeat domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3192 - microRNA 3192 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3F - RNA polymerase III subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282526.1 | 348 | Silent Mutation | AAA,AAG | K,K 36 | NP_001269455.1 | |
NM_006466.3 | 348 | Silent Mutation | AAA,AAG | K,K 77 | NP_006457.2 | |
XM_011529141.1 | 348 | UTR 5 | XP_011527443.1 | |||
XM_017027596.1 | 348 | Silent Mutation | AAA,AAG | K,K 77 | XP_016883085.1 |