Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGCAGCCCTGCTGGGGATGTT[C/G]AGTTGCTGCAAGGTTCATGCTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610106 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBNL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DBNL - drebrin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014436.2 | Intron | NP_001014436.1 | ||||
NM_001122956.1 | Intron | NP_001116428.1 | ||||
NM_001284313.1 | Intron | NP_001271242.1 | ||||
NM_001284315.1 | Intron | NP_001271244.1 | ||||
NM_014063.6 | Intron | NP_054782.2 | ||||
XM_011515325.1 | Intron | XP_011513627.1 |
LINC00957 - long intergenic non-protein coding RNA 957 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6837 - microRNA 6837 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASA4CP - RAS p21 protein activator 4C, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |