Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609526 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCTD19 - potassium channel tetramerization domain containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100915.1 | Intron | NP_001094385.1 |
PLEKHG4 - pleckstrin homology and RhoGEF domain containing G4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129727.2 | Intron | NP_001123199.1 | ||||
NM_001129728.1 | Intron | NP_001123200.1 | ||||
NM_001129729.2 | Intron | NP_001123201.1 | ||||
NM_001129731.2 | Intron | NP_001123203.1 | ||||
XM_011522985.2 | Intron | XP_011521287.1 | ||||
XM_011522986.2 | Intron | XP_011521288.1 | ||||
XM_011522987.2 | Intron | XP_011521289.1 | ||||
XM_011522988.2 | Intron | XP_011521290.1 |
Set Membership: |
HapMap |