Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACTCTGAGGTGTGGAAAATCTG[C/T]ATCACGCCCCAACTAAACGTCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601323 MIM: 602309 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUCB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUCB1 - nucleobindin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006184.5 | Intron | NP_006175.2 | ||||
XM_017026845.1 | Intron | XP_016882334.1 |
NUCB1-AS1 - NUCB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TULP2 - tubby like protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003323.2 | Intron | NP_003314.2 | ||||
XM_006723347.3 | Intron | XP_006723410.1 | ||||
XM_006723348.3 | Intron | XP_006723411.1 | ||||
XM_011527252.2 | Intron | XP_011525554.1 | ||||
XM_011527253.2 | Intron | XP_011525555.1 | ||||
XM_011527254.2 | Intron | XP_011525556.1 | ||||
XM_011527255.2 | Intron | XP_011525557.1 | ||||
XM_011527256.2 | Intron | XP_011525558.1 |
Set Membership: |
HapMap |