Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR1E2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OR1E2 - olfactory receptor family 1 subfamily E member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA22 - spermatogenesis associated 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170695.1 | 1237 | Silent Mutation | CCA,CCG | P,P 333 | NP_001164166.1 | |
NM_001170696.1 | 1237 | Silent Mutation | CCA,CCG | P,P 290 | NP_001164167.1 | |
NM_001170697.1 | 1237 | Silent Mutation | CCA,CCG | P,P 333 | NP_001164168.1 | |
NM_001170698.1 | 1237 | Silent Mutation | CCA,CCG | P,P 333 | NP_001164169.1 | |
NM_001170699.1 | 1237 | UTR 3 | NP_001164170.1 | |||
NM_001321336.1 | 1237 | Silent Mutation | CCA,CCG | P,P 317 | NP_001308265.1 | |
NM_001321337.1 | 1237 | Silent Mutation | CCA,CCG | P,P 333 | NP_001308266.1 | |
NM_032598.4 | 1237 | Silent Mutation | CCA,CCG | P,P 333 | NP_115987.2 |