Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAATAGCTGTTTAAAACTTTTT[A/T]AAAAACGTAAATCTTATCAGCTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IQUB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQUB - IQ motif and ubiquitin domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282855.1 | Intron | NP_001269784.1 | ||||
NM_001321293.1 | Intron | NP_001308222.1 | ||||
NM_178827.4 | Intron | NP_849149.3 | ||||
XM_005250161.3 | Intron | XP_005250218.1 | ||||
XM_005250162.4 | Intron | XP_005250219.1 | ||||
XM_011515833.2 | Intron | XP_011514135.1 | ||||
XM_011515834.2 | Intron | XP_011514136.1 | ||||
XM_017011771.1 | Intron | XP_016867260.1 | ||||
XM_017011772.1 | Intron | XP_016867261.1 |