Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606848 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NEK7 - NIMA related kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133494.2 | Intron | NP_598001.1 | ||||
XM_011509209.1 | Intron | XP_011507511.1 | ||||
XM_017000344.1 | Intron | XP_016855833.1 | ||||
XM_017000345.1 | Intron | XP_016855834.1 | ||||
XM_017000346.1 | Intron | XP_016855835.1 | ||||
XM_017000347.1 | Intron | XP_016855836.1 | ||||
XM_017000348.1 | Intron | XP_016855837.1 |
Set Membership: |
HapMap |