Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610510 MIM: 613448 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BORA - bora, aurora kinase A activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286746.1 | Intron | NP_001273675.1 | ||||
NM_001286747.1 | Intron | NP_001273676.1 | ||||
NM_024808.3 | Intron | NP_079084.3 | ||||
XM_006719868.3 | Intron | XP_006719931.2 |
MZT1 - mitotic spindle organizing protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |