Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104220 | ||||||||||||||||||||
Literature Links: |
ADRA1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADRA1B - adrenoceptor alpha 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000679.3 | 325 | Missense Mutation | GGG,GTG | G,V 51 | NP_000670.1 | |
XM_005265818.3 | 325 | Missense Mutation | GGG,GTG | G,V 51 | XP_005265875.1 | |
XM_005265819.2 | 325 | Missense Mutation | GGG,GTG | G,V 51 | XP_005265876.1 | |
XM_006714821.3 | 325 | Missense Mutation | GGG,GTG | G,V 51 | XP_006714884.1 | |
XM_011534435.1 | 325 | Missense Mutation | GGG,GTG | G,V 51 | XP_011532737.1 | |
XM_011534437.2 | 325 | Missense Mutation | GGG,GTG | G,V 51 | XP_011532739.1 | |
XM_011534438.2 | 325 | Missense Mutation | GGG,GTG | G,V 51 | XP_011532740.1 |