Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP64 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFP64 - ZFP64 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319146.1 | Intron | NP_001306075.1 | ||||
NM_018197.2 | Intron | NP_060667.2 | ||||
NM_022088.4 | Intron | NP_071371.3 | ||||
NM_199426.1 | Intron | NP_955458.1 | ||||
NM_199427.2 | Intron | NP_955459.2 | ||||
XM_005260449.1 | Intron | XP_005260506.1 | ||||
XM_006723822.1 | Intron | XP_006723885.1 | ||||
XM_011528900.1 | Intron | XP_011527202.1 | ||||
XM_017027945.1 | Intron | XP_016883434.1 | ||||
XM_017027946.1 | Intron | XP_016883435.1 | ||||
XM_017027947.1 | Intron | XP_016883436.1 | ||||
XM_017027948.1 | Intron | XP_016883437.1 | ||||
XM_017027949.1 | Intron | XP_016883438.1 |
Set Membership: |
HapMap |