Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610372 MIM: 605521 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC2A12 - solute carrier family 2 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145176.2 | 4954 | UTR 3 | NP_660159.1 | |||
XM_006715349.3 | 4954 | Intron | XP_006715412.1 | |||
XM_017010311.1 | 4954 | Intron | XP_016865800.1 |
TBPL1 - TATA-box binding protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253676.1 | 4954 | Intron | NP_001240605.1 | |||
NM_004865.3 | 4954 | Intron | NP_004856.1 | |||
XM_017011513.1 | 4954 | Intron | XP_016867002.1 |