Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGATATTTACTTACTATACCACA[A/T]GAAATAATATAAGGAAGGAGAAAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611673 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRMT1L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRMT1L - tRNA methyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202423.1 | Intron | NP_001189352.1 | ||||
NM_030934.4 | Intron | NP_112196.3 | ||||
XM_011510028.2 | Intron | XP_011508330.1 | ||||
XM_017002419.1 | Intron | XP_016857908.1 |
Set Membership: |
HapMap |