Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147460 | ||||||||||||||||||||
Literature Links: |
SOD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SOD2 - superoxide dismutase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000636.3 | 871 | UTR 3 | NP_000627.2 | |||
NM_001024465.2 | 871 | Intron | NP_001019636.1 | |||
NM_001024466.2 | 871 | Intron | NP_001019637.1 | |||
NM_001322814.1 | 871 | UTR 3 | NP_001309743.1 | |||
NM_001322815.1 | 871 | UTR 3 | NP_001309744.1 | |||
NM_001322816.1 | 871 | Intron | NP_001309745.1 | |||
NM_001322817.1 | 871 | Intron | NP_001309746.1 | |||
NM_001322819.1 | 871 | UTR 3 | NP_001309748.1 | |||
NM_001322820.1 | 871 | UTR 3 | NP_001309749.1 |