Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGAATCCAAGTATCTCAGCAC[A/G]TGGACCCCAGAGCTTTTTTTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOPL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPOPL - speckle type BTB/POZ protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001664.2 | Intron | NP_001001664.1 | ||||
XM_005263655.4 | Intron | XP_005263712.1 | ||||
XM_005263656.4 | Intron | XP_005263713.1 | ||||
XM_005263657.4 | Intron | XP_005263714.1 |
Set Membership: |
HapMap |