Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAACTAGCAGTGCTGTGGGAC[A/G]ACCTCATGAAGGGACCCAGTGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603447 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HRK - harakiri, BCL2 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003806.2 | 5528 | Intron | NP_003797.1 | |||
XM_017020146.1 | 5528 | UTR 3 | XP_016875635.1 |
RNFT2 - ring finger protein, transmembrane 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |