Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610919 MIM: 606472 | ||||||||||||||||||||
Literature Links: |
MTG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTG2 - mitochondrial ribosome associated GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015666.3 | Intron | NP_056481.1 | ||||
XM_005260393.1 | Intron | XP_005260450.1 | ||||
XM_006723774.3 | Intron | XP_006723837.1 | ||||
XM_011528776.1 | Intron | XP_011527078.1 | ||||
XM_017027805.1 | Intron | XP_016883294.1 | ||||
XM_017027806.1 | Intron | XP_016883295.1 | ||||
XM_017027807.1 | Intron | XP_016883296.1 | ||||
XM_017027808.1 | Intron | XP_016883297.1 |
SS18L1 - SS18L1, nBAF chromatin remodeling complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |