Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTAAAAACAAAGTTGTTTCCC[A/G]TATTTCCACCTTCGTTCCTAGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600421 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLRA3 - glycine receptor alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042543.2 | 2605 | UTR 3 | NP_001036008.1 | |||
NM_006529.3 | 2605 | UTR 3 | NP_006520.2 | |||
XM_011532267.2 | 2605 | Intron | XP_011530569.1 | |||
XM_011532268.2 | 2605 | UTR 3 | XP_011530570.1 | |||
XM_017008630.1 | 2605 | UTR 3 | XP_016864119.1 |
Set Membership: |
HapMap |