Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCCTGAACAATTAGAAGAATT[C/T]GTATAAGTAGAGAGCAAGGTAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603330 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAH9 - dynein axonemal heavy chain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001372.3 | Intron | NP_001363.2 | ||||
NM_004662.2 | Intron | NP_004653.2 | ||||
XM_011523703.1 | Intron | XP_011522005.1 | ||||
XM_017024292.1 | Intron | XP_016879781.1 | ||||
XM_017024293.1 | Intron | XP_016879782.1 | ||||
XM_017024294.1 | Intron | XP_016879783.1 | ||||
XM_017024295.1 | Intron | XP_016879784.1 |
Set Membership: |
HapMap |