Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CASS4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CASS4 - Cas scaffolding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164114.1 | 2511 | Missense Mutation | AAT,ACT | N,T 575 | NP_001157586.1 | |
NM_001164115.1 | 2511 | Intron | NP_001157587.1 | |||
NM_001164116.1 | 2511 | Missense Mutation | AAT,ACT | N,T 629 | NP_001157588.1 | |
NM_020356.3 | 2511 | Missense Mutation | AAT,ACT | N,T 629 | NP_065089.2 | |
XM_006723831.3 | 2511 | Missense Mutation | AAT,ACT | N,T 575 | XP_006723894.1 |