Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGATGAGACTGTGGGTTGGTTT[C/T]CCTTGAAGACAATGTGGGTGTGCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616380 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMB4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LAMB4 - laminin subunit beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318046.1 | Intron | NP_001304975.1 | ||||
NM_001318047.1 | Intron | NP_001304976.1 | ||||
NM_001318048.1 | Intron | NP_001304977.1 | ||||
NM_007356.2 | Intron | NP_031382.2 | ||||
XM_011515975.1 | Intron | XP_011514277.1 | ||||
XM_011515978.1 | Intron | XP_011514280.1 | ||||
XM_011515979.2 | Intron | XP_011514281.1 | ||||
XM_011515980.2 | Intron | XP_011514282.1 | ||||
XM_017011879.1 | Intron | XP_016867368.1 | ||||
XM_017011880.1 | Intron | XP_016867369.1 |
Set Membership: |
HapMap |