Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTAGAAGTATATACACATTTCC[A/G]TATTCCCCTCATTTTTCCCTCTCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606531 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C5orf46 - chromosome 5 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206966.2 | 1149 | Intron | NP_996849.2 | |||
XM_005268446.3 | 1149 | UTR 3 | XP_005268503.2 | |||
XM_017009459.1 | 1149 | Intron | XP_016864948.1 |
SCGB3A2 - secretoglobin family 3A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |